4-(1,3,4-oxadiazol-2-ylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

46533-500MG 46533-1G 46533-5G
77335-496EA 356.38 USD
77335-496 77350-774 77375-012
4-(1,3,4-oxadiazol-2-ylphenyl)boronic acid ≥97%
4-(1,3,4-oxadiazol-2-ylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BN₂O₃
MW: 189.964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11865276
CAS Number: 276694-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR