2-Fluoro-3-iodo-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46529-500MG 46529-1G 46529-5G 46529-25G
77327-188EA 208.06 USD
77327-188 77350-772 77363-460 77386-094
2-Fluoro-3-iodo-4-methoxybenzoic acid ≥97%
2-Fluoro-3-iodo-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₃
MW: 296.034 g/mol
Storage Temperature: Ambient
CAS Number: 2385874-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR