2-Bromo-1-chloro-4-(cyclopropylmethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

46517-500MG 46517-1G 46517-5G
77327-186EA 325.48 USD
77327-186 77350-770 77375-010
2-Bromo-1-chloro-4-(cyclopropylmethyl)benzene ≥95%
2-Bromo-1-chloro-4-(cyclopropylmethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrCl
MW: 245.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092491
CAS Number: 1369951-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR