1-Bromo-4-(cyclopropylmethyl)-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46516-500MG 46516-1G 46516-5G
77335-802EA 366.68 USD
77335-802 77350-768 77375-008
1-Bromo-4-(cyclopropylmethyl)-2-fluorobenzene ≥95%
1-Bromo-4-(cyclopropylmethyl)-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD29091144
CAS Number: 1369919-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR