1-Bromo-2-chloro-4-(cyclopropylmethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

46515-500MG 46515-1G 46515-5G
77327-184EA 325.48 USD
77327-184 77350-766 77375-006
1-Bromo-2-chloro-4-(cyclopropylmethyl)benzene ≥97%
1-Bromo-2-chloro-4-(cyclopropylmethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrCl
MW: 245.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29092492
CAS Number: 1369900-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR