4-(Cyclopropylmethyl)-1,2-difluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46513-500MG 46513-1G 46513-5G 46513-25G
77327-182EA 263.68 USD
77327-182 77350-764 77363-458 77386-092
4-(Cyclopropylmethyl)-1,2-difluorobenzene ≥95%
4-(Cyclopropylmethyl)-1,2-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂
MW: 168.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091045
CAS Number: 1369849-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR