2-Bromo-4-(cyclopropylmethyl)-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

46511-500MG 46511-1G 46511-25G 46511-5G
77327-180EA 299.73 USD
77327-180 77350-762 77386-090 77375-004
2-Bromo-4-(cyclopropylmethyl)-1-fluorobenzene ≥97%
2-Bromo-4-(cyclopropylmethyl)-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD29091143
CAS Number: 1369790-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR