2-Chloro-4-(cyclopropylmethyl)-1-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46510-500MG 46510-1G 46510-5G 46510-25G
77327-178EA 263.68 USD
77327-178 77350-760 77363-456 77386-088
2-Chloro-4-(cyclopropylmethyl)-1-fluorobenzene ≥95%
2-Chloro-4-(cyclopropylmethyl)-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClF
MW: 184.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091087
CAS Number: 1369774-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR