2-Bromo-4-(cyclopropylmethyl)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

46501-500MG 46501-1G 46501-5G
77336-370EA 387.28 USD
77336-370 77350-758 77375-002
2-Bromo-4-(cyclopropylmethyl)-1-methylbenzene ≥97%
2-Bromo-4-(cyclopropylmethyl)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃Br
MW: 225.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094596
CAS Number: 1369849-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR