1-Bromo-3-(cyclopropylmethyl)-5-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

46494-500MG 46494-1G 46494-5G
77327-176EA 325.48 USD
77327-176 77350-756 77375-000
1-Bromo-3-(cyclopropylmethyl)-5-fluorobenzene ≥95%
1-Bromo-3-(cyclopropylmethyl)-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD29091147
CAS Number: 1369880-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR