1-Bromo-4-(cyclopropylmethyl)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

46464-500MG 46464-1G 46464-5G
77327-172EA 346.08 USD
77327-172 77350-752 77374-996
1-Bromo-4-(cyclopropylmethyl)-2-methylbenzene ≥95%
1-Bromo-4-(cyclopropylmethyl)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃Br
MW: 225.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094595
CAS Number: 1369916-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR