(E)-N'-((5-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46462-1G 46462-5G 46462-25G
77341-134EA 72.1 USD
77341-134 77363-454 77374-994
(E)-N'-((5-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%
(E)-N'-((5-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD32707176
CAS Number: 2415435-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR