(E)-N'-((6-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46461-1G 46461-5G 46461-25G
77341-132EA 72.1 USD
77341-132 77363-452 77374-992
(E)-N'-((6-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥97%
(E)-N'-((6-Bromopyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD32707175
CAS Number: 2432855-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR