3-Bromo-5-(cyclopropylmethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

46374-500MG 46374-1G 46374-5G 46374-25G
77327-168EA 189.52 USD
77327-168 77350-748 77363-448 77386-084
3-Bromo-5-(cyclopropylmethyl)pyridine ≥97%
3-Bromo-5-(cyclopropylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
MW: 212.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564099
CAS Number: 2412764-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR