(2-Ethoxy-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

46367-500MG 46367-1G 46367-5G 46367-25G
77327-166EA 202.91 USD
77327-166 77350-746 77363-446 77386-082
(2-Ethoxy-4-methylphenyl)methanol ≥97%
(2-Ethoxy-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707174
CAS Number: 2432849-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR