Methyl 2-ethoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

46366-500MG 46366-1G 46366-5G 46366-25G
77327-164EA 226.6 USD
77327-164 77350-744 77363-444 77386-080
Methyl 2-ethoxy-4-methylbenzoate ≥97%
Methyl 2-ethoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403839
CAS Number: 1052647-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR