Ethyl 2,3-dichloro-4-methylbenzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
46363-500MG
46363-1G
46363-5G
77339-114EA
552.08
USD
77339-114
77350-742
77374-988
Ethyl 2,3-dichloro-4-methylbenzoate ≥95%
Ethyl 2,3-dichloro-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀Cl₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD28786803 CAS Number: 1803844-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...