Methyl 3-fluoro-4-propoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

46341-500MG 46341-1G 46341-5G 46341-25G
77327-158EA 281.19 USD
77327-158 77350-738 77363-438 77386-076
Methyl 3-fluoro-4-propoxybenzoate ≥95%
Methyl 3-fluoro-4-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403837
CAS Number: 2635937-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR