Ethyl 6-bromo-2-fluoro-3-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46338-500MG 46338-1G 46338-5G 46338-25G
77327-154EA 108.15 USD
77327-154 77343-216 77363-434 77374-984
Ethyl 6-bromo-2-fluoro-3-propoxybenzoate ≥97%
Ethyl 6-bromo-2-fluoro-3-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34564098
CAS Number: 2734772-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR