Methyl 6-bromo-2-fluoro-3-propoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

46337-500MG 46337-1G 46337-5G 46337-25G
77327-152EA 108.15 USD
77327-152 77343-214 77363-432 77374-982
Methyl 6-bromo-2-fluoro-3-propoxybenzoate ≥95%
Methyl 6-bromo-2-fluoro-3-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34564097
CAS Number: 2734772-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR