6-Bromo-2-fluoro-3-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46336-1G 46336-5G 46336-25G
77341-316EA 82.4 USD
77341-316 77363-430 77374-980
6-Bromo-2-fluoro-3-propoxybenzoic acid ≥97%
6-Bromo-2-fluoro-3-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD33403836
CAS Number: 2624417-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR