(6-Bromo-2-fluoro-3-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

46335-500MG 46335-1G 46335-5G 46335-25G
77327-150EA 136.99 USD
77327-150 77344-586 77363-428 77386-072
(6-Bromo-2-fluoro-3-propoxyphenyl)methanol ≥97%
(6-Bromo-2-fluoro-3-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34178523
CAS Number: 2643368-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR