3,4-Difluoro-2-methoxy-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

46314-500MG 46314-1G 46314-5G
77336-368EA 387.28 USD
77336-368 77350-726 77374-970
3,4-Difluoro-2-methoxy-5-methylbenzaldehyde ≥95%
3,4-Difluoro-2-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707171
CAS Number: 2484889-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR