2-(3,4-Difluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

46310-500MG 46310-1G 46310-5G 46310-25G
77327-140EA 201.88 USD
77327-140 77350-722 77363-422 77386-066
2-(3,4-Difluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%
2-(3,4-Difluoro-2-methoxyphenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902699
CAS Number: 2586126-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR