3,5-Difluoro-4-isobutoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46302-500MG 46302-1G 46302-5G 46302-25G
77327-134EA 202.91 USD
77327-134 77350-716 77363-418 77386-062
3,5-Difluoro-4-isobutoxybenzoic acid ≥97%
3,5-Difluoro-4-isobutoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18285386
CAS Number: 1343670-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR