3,5-Difluoro-4-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46300-500MG 46300-1G 46300-5G 46300-25G
77327-132EA 237.93 USD
77327-132 77350-714 77363-416 77386-060
3,5-Difluoro-4-isobutoxybenzaldehyde ≥97%
3,5-Difluoro-4-isobutoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18426710
CAS Number: 1443343-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR