N-(5-Chloropyridin-2-yl)acetamide ≥97%

Supplier: AOBChem USA

Warning

46291-500MG 46291-1G 46291-5G 46291-25G
77327-128EA 160.68 USD
77327-128 77345-786 77363-410 77386-056
N-(5-Chloropyridin-2-yl)acetamide ≥97%
N-(5-Chloropyridin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02577036
CAS Number: 45965-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR