4'-Methyl-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

46284-1G 46284-5G 46284-25G
77341-522EA 91.67 USD
77341-522 77363-408 77374-962
4'-Methyl-1,1':3',1''-terphenyl ≥97%
4'-Methyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₉H₁₆
Storage Temperature: Ambient
MDL Number: MFCD32707169
CAS Number: 10468-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR