1-(3-Bromo-2,6-difluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

46236-500MG 46236-1G 46236-5G 46236-25G
77327-120EA 184.37 USD
77327-120 77347-194 77363-402 77386-048
1-(3-Bromo-2,6-difluorophenyl)ethanone ≥97%
1-(3-Bromo-2,6-difluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD24369744
CAS Number: 1387634-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR