1-(2-Bromo-3-fluoropyridin-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

46227-500MG 46227-1G 46227-5G
77339-404EA 586.07 USD
77339-404 77350-706 77374-958
1-(2-Bromo-3-fluoropyridin-4-yl)ethanone ≥97%
1-(2-Bromo-3-fluoropyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34181041
CAS Number: 2734772-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR