3-Fluoro-4-hydroxy-5-(trifluoromethyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
46218-500MG
46218-1G
46218-25G
46218-5G
77327-116EA
298.7
USD
77327-116
77350-702
77386-044
77374-952
3-Fluoro-4-hydroxy-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Fluoro-4-hydroxy-5-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BF₄O₃ MW: 223.917 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403828 CAS Number: 2260683-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...