4-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

46214-500MG 46214-1G 46214-5G
77336-366EA 387.28 USD
77336-366 77350-698 77374-948
4-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
4-Ethoxy-3-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403826
CAS Number: 2624417-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR