3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46210-500MG 46210-1G 46210-5G
77338-212EA 469.68 USD
77338-212 77350-696 77374-946
3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Fluoro-4-isopropoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403824
CAS Number: 2624417-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR