4-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

46202-500MG 46202-1G 46202-5G 46202-25G
77327-112EA 255.44 USD
77327-112 77350-686 77363-398 77386-042
4-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
4-(Benzyloxy)-3-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀F₄O₃
MW: 314.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403819
CAS Number: 2635937-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR