2-(Benzyloxy)-5-bromo-1-fluoro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

46199-500MG 46199-1G 46199-5G 46199-25G
77327-106EA 119.48 USD
77327-106 77343-600 77363-396 77374-932
2-(Benzyloxy)-5-bromo-1-fluoro-3-(trifluoromethyl)benzene ≥97%
2-(Benzyloxy)-5-bromo-1-fluoro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902698
CAS Number: 2586125-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR