N-(4-Bromo-2-chloro-6-methylphenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

46198-500MG 46198-1G 46198-5G 46198-25G
77327-104EA 231.75 USD
77327-104 77350-680 77363-394 77386-038
N-(4-Bromo-2-chloro-6-methylphenyl)acetamide ≥95%
N-(4-Bromo-2-chloro-6-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉BrClNO
MW: 262.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30275796
CAS Number: 1863924-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR