Methyl 3,5-difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
46195-500MG
46195-1G
46195-25G
46195-5G
77327-098EA
305.91
USD
77327-098
77350-674
77386-032
77374-928
Methyl 3,5-difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylate ≥97%
Methyl 3,5-difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₂F₂O₂ MW: 262.251 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403815 CAS Number: 2635937-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...