3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

46191-500MG 46191-1G 46191-25G 46191-5G
77327-094EA 304.88 USD
77327-094 77350-668 77386-028 77374-924
3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564094
CAS Number: 2734775-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR