3-Bromo-5-butoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46183-500MG 46183-1G 46183-25G 46183-5G
77327-092EA 304.88 USD
77327-092 77350-666 77386-026 77374-922
3-Bromo-5-butoxybenzaldehyde ≥97%
3-Bromo-5-butoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707165
CAS Number: 1973419-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR