(4-Methoxy-3-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

46181-500MG 46181-1G 46181-5G
77335-934EA 369.77 USD
77335-934 77350-664 77374-920
(4-Methoxy-3-(methylthio)phenyl)methanol ≥97%
(4-Methoxy-3-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD29934665
CAS Number: 96495-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR