Ethyl 6-ethoxypicolinate ≥97%

Supplier: AOBChem USA

Warning

46172-500MG 46172-1G 46172-5G 46172-25G
77327-090EA 126.69 USD
77327-090 77344-208 77363-388 77374-918
Ethyl 6-ethoxypicolinate ≥97%
Ethyl 6-ethoxypicolinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977443
CAS Number: 890655-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR