3-Bromo-4-(methoxymethyl)thiophene ≥97%

Supplier: AOBChem USA

Warning

46163-500MG 46163-1G 46163-5G 46163-25G
77327-084EA 241.02 USD
77327-084 77350-662 77363-382 77386-022
3-Bromo-4-(methoxymethyl)thiophene ≥97%
3-Bromo-4-(methoxymethyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₇BrOS
MW: 207.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707164
CAS Number: 2432848-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR