Ethyl 2-fluoro-5-propoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46158-500MG 46158-1G 46158-5G 46158-25G
77327-080EA 202.91 USD
77327-080 77350-658 77363-378 77386-018
Ethyl 2-fluoro-5-propoxybenzoate ≥97%
Ethyl 2-fluoro-5-propoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD33403814
CAS Number: 2635937-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR