2-Fluoro-5-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46156-500MG 46156-1G 46156-5G 46156-25G
77327-076EA 120.51 USD
77327-076 77343-708 77363-374 77374-914
2-Fluoro-5-propoxybenzoic acid ≥97%
2-Fluoro-5-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403813
CAS Number: 2154990-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR