Methyl 5-(benzyloxy)-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

46149-500MG 46149-1G 46149-25G 46149-5G
77327-074EA 304.88 USD
77327-074 77350-656 77386-014 77374-908
Methyl 5-(benzyloxy)-2-fluorobenzoate ≥95%
Methyl 5-(benzyloxy)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902696
CAS Number: 2586126-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR