5-(Benzyloxy)-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

46147-500MG 46147-1G 46147-5G 46147-25G
77327-070EA 263.68 USD
77327-070 77350-652 77363-368 77386-010
5-(Benzyloxy)-2-fluorobenzoic acid ≥97%
5-(Benzyloxy)-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₃
MW: 246.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902694
CAS Number: 1148146-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR