(5-Ethoxy-2-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

46141-500MG 46141-1G 46141-5G 46141-25G
77327-062EA 210.12 USD
77327-062 77350-646 77363-364 77386-004
(5-Ethoxy-2-fluorophenyl)methanol ≥95%
(5-Ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD32902691
CAS Number: 2586126-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR