3-(Benzyloxy)-2-bromobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

46121-500MG 46121-1G 46121-5G 46121-25G
77327-058EA 151.41 USD
77327-058 77345-388 77363-360 77386-000
3-(Benzyloxy)-2-bromobenzaldehyde ≥95%
3-(Benzyloxy)-2-bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD30575616
CAS Number: 158584-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR