Methyl 3-chloro-5-fluoro-4-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46103-500MG 46103-1G 46103-5G
77327-056EA 347.11 USD
77327-056 77350-640 77374-896
Methyl 3-chloro-5-fluoro-4-isopropoxybenzoate ≥97%
Methyl 3-chloro-5-fluoro-4-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO₃
MW: 246.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707156
CAS Number: 2432848-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR