(3-Chloro-5-fluoro-4-isopropoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

46100-500MG 46100-1G 46100-5G 46100-25G
77327-054EA 243.08 USD
77327-054 77350-638 77363-358 77385-998
(3-Chloro-5-fluoro-4-isopropoxyphenyl)methanol ≥95%
(3-Chloro-5-fluoro-4-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO₂
MW: 218.652 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707155
CAS Number: 2432848-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR